Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmtc4
Entrez Gene ID
70551
Gene Name
transmembrane and tetratricopeptide repeat containing 4
Gene Aliases
4930403J22Rik, 5730419O14Rik
Chromosome Location
Chr.14: 122918971 - 122983283 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028651.2 NP_082927.1 4 - 5 640 78
GenBank mRNA AK028333.1 - 4 - 5 621 78
AK033947.1 - 4 - 5 641 78
AK036741.1 - 4 - 5 597 78
AK039093.1 - 4 - 5 585 78
AK043293.1 - 4 - 5 667 78
AK044391.1 - 4 - 5 596 78
AK045271.1 - 4 - 5 602 78
AK049748.1 - 5 - 6 781 78
AK082526.1 - 3 - 4 468 78

View Full Product Details