Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vprbp
Entrez Gene ID
321006
Gene Name
Vpr (HIV-1) binding protein
Gene Aliases
AI447437, B930007L02Rik, mKIAA0800
Chromosome Location
Chr.9: 106814172 - 106880992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015507.2 NP_001015507.1 21 - 22 4228 73
GenBank mRNA AK029372.1 - 4 - 5 320 73
AK122372.1 - 21 - 22 4191 73
BC065119.1 - 21 - 22 4228 73

View Full Product Details