Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sulf1
Entrez Gene ID
240725
Gene Name
sulfatase 1
Gene Aliases
AI467640, AW121680, mKIAA1077
Chromosome Location
Chr.1: 12691695 - 12861192 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198565.1 NP_001185494.1 15 - 16 2421 87
NM_001198566.1 NP_001185495.1 14 - 15 2326 87
NM_172294.1 NP_758498.1 15 - 16 2405 87
GenBank mRNA AK028285.1 - 14 - 15 2225 87
AK045002.1 - 15 - 16 2320 87
AK078756.1 - 15 - 16 2410 87
AK129278.1 - 14 - 15 2227 87
AY101178.1 - 15 - 16 2430 87
BC034547.1 - 15 - 16 2405 87
BC049276.1 - 15 - 16 2405 87

View Full Product Details