Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgef33
Entrez Gene ID
381112
Gene Name
Rho guanine nucleotide exchange factor (GEF) 33
Gene Aliases
Gm941
Chromosome Location
Chr.17: 80306930 - 80427079 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145452.1 NP_001138924.1 7 - 8 786 103
GenBank mRNA AK053944.1 - 3 - 4 420 103
CK793330.1 - 4 - 5 464 103

View Full Product Details