Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eml4
Entrez Gene ID
78798
Gene Name
echinoderm microtubule associated protein like 4
Gene Aliases
4930443C24Rik, AI644019
Chromosome Location
Chr.17: 83350931 - 83480361 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114361.1 NP_001107833.1 21 - 22 2560 69
NM_001114362.1 NP_001107834.1 19 - 20 2353 69
NM_001286567.1 NP_001273496.1 21 - 22 2463 69
NM_199466.3 NP_955760.3 18 - 19 2224 69
GenBank mRNA AK019611.1 - 2 - 3 182 69
AK077527.1 - 13 - 14 1389 69
AK144604.1 - 21 - 22 2561 69
BC032258.1 - 7 - 8 708 69
BC065178.1 - 21 - 22 2401 69
BC066099.1 - 18 - 19 2155 69
BC067011.1 - 18 - 19 2173 69
BC070427.1 - 19 - 20 2293 69
BC079619.1 - 18 - 19 2155 69
DQ230326.1 - 20 - 21 2243 69

View Full Product Details