The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161837.1 | NP_001155309.1 | 4 - 5 | 510 | 127 |
NM_001161838.1 | NP_001155310.1 | 1 - 2 | 467 | 127 | |
NM_001161839.1 | NP_001155311.1 | 2 - 3 | 158 | 127 | |
NM_001161840.1 | NP_001155312.1 | 2 - 3 | 350 | 127 | |
NM_011217.2 | NP_035347.1 | 5 - 6 | 1116 | 127 | |
GenBank mRNA | AF041866.1 | - | 2 - 3 | 275 | 127 |
AF129509.1 | - | 1 - 2 | 392 | 127 | |
AK028054.1 | - | 5 - 6 | 1117 | 127 | |
BC119230.1 | - | 5 - 6 | 1049 | 127 | |
BC120547.1 | - | 5 - 6 | 1049 | 127 | |
D31898.1 | - | 5 - 6 | 1079 | 127 | |
DV657487.1 | - | 2 - 3 | 350 | 127 | |
Z30313.1 | - | 4 - 5 | 505 | 127 |