Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dgat2l6
Entrez Gene ID
668257
Gene Name
diacylglycerol O-acyltransferase 2-like 6
Gene Aliases
9930021F22Rik
Chromosome Location
Chr.X: 100516094 - 100546108 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114084.1 NP_001107556.1 5 - 6 731 99

View Full Product Details