Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Safb
Entrez Gene ID
224903
Gene Name
scaffold attachment factor B
Gene Aliases
3110021E02Rik, 5330423C17Rik, AU018122, D18386, E130307D12, HAP, HET, SAF-B1, SAFB1
Chromosome Location
Chr.17: 56584760 - 56606294 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163300.1 NP_001156772.1 16 - 17 2541 67
GenBank mRNA AK087504.1 - 9 - 10 3001 67

View Full Product Details