Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxo11
Entrez Gene ID
225055
Gene Name
F-box protein 11
Gene Aliases
C80048, Jf
Chromosome Location
Chr.17: 87990859 - 88065285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081034.1 NP_001074503.1 10 - 11 1306 94
GenBank mRNA BC055343.1 - 1 - 2 48 94
BC110686.1 - 10 - 11 1045 94
BC117884.1 - 10 - 11 1052 94
BC117885.1 - 10 - 11 1056 94
BC128479.1 - 10 - 11 1056 94

View Full Product Details