Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxo11
Entrez Gene ID
225055
Gene Name
F-box protein 11
Gene Aliases
C80048, Jf
Chromosome Location
Chr.17: 87990859 - 88065285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081034.1 NP_001074503.1 21 - 22 2597 69
GenBank mRNA AK132635.1 - 5 - 6 573 69
BC027802.1 - 10 - 11 1174 69
BC049946.1 - 5 - 6 525 69
BC055343.1 - 12 - 13 1339 69
BC110686.1 - 21 - 22 2336 69
BC117884.1 - 21 - 22 2343 69
BC117885.1 - 21 - 22 2347 69
BC128479.1 - 21 - 22 2347 69

View Full Product Details