Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vit
Entrez Gene ID
74199
Gene Name
vitrin
Gene Aliases
1700052E02Rik, 1700110E08Rik, 2810429K11Rik
Chromosome Location
Chr.17: 78508051 - 78627409 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197028.1 NP_001183957.1 6 - 7 889 57
NM_028813.2 NP_083089.1 7 - 8 955 57
GenBank mRNA AK013193.1 - 7 - 8 866 57
AK051606.1 - 6 - 7 780 57
BC034120.1 - 7 - 8 878 57

View Full Product Details