The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164074.1 | NP_001157546.1 | 2 - 3 | 304 | 83 |
NM_001164075.1 | NP_001157547.1 | 2 - 3 | 644 | 83 | |
NM_001164076.1 | NP_001157548.1 | 2 - 3 | 370 | 83 | |
NM_001164077.1 | NP_001157549.1 | 1 - 2 | 598 | 83 | |
NM_009372.3 | NP_033398.2 | 2 - 3 | 558 | 83 | |
GenBank mRNA | AK008909.1 | - | 2 - 3 | 560 | 83 |
AK160193.1 | - | 2 - 3 | 263 | 83 | |
BC005724.1 | - | 2 - 3 | 557 | 83 | |
BC012700.1 | - | 2 - 3 | 655 | 83 | |
BY096128.1 | - | 2 - 3 | 308 | 83 | |
CD563532.1 | - | 2 - 3 | 474 | 83 | |
X89749.1 | - | 2 - 3 | 514 | 83 |