Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nicn1
Entrez Gene ID
66257
Gene Name
nicolin 1
Gene Aliases
1500032A17Rik
Chromosome Location
Chr.9: 108290457 - 108296496 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025449.3 NP_079725.1 3 - 4 501 78
GenBank mRNA AJ299741.1 - 3 - 4 493 78
AK002789.1 - 3 - 4 507 78
AK013602.1 - 3 - 4 529 78
AK075732.1 - 3 - 4 509 78
AK146890.1 - 3 - 4 511 78
AK169135.1 - 3 - 4 511 78
BC024050.1 - 3 - 4 501 78

View Full Product Details