Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nicn1
Entrez Gene ID
66257
Gene Name
nicolin 1
Gene Aliases
1500032A17Rik
Chromosome Location
Chr.9: 108290457 - 108296496 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025449.3 NP_079725.1 4 - 5 565 68
GenBank mRNA AJ299741.1 - 4 - 5 557 68
AK002789.1 - 4 - 5 571 68
AK013602.1 - 4 - 5 593 68
AK075732.1 - 4 - 5 573 68
BC024050.1 - 4 - 5 565 68

View Full Product Details