Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tyms
- Entrez Gene ID
-
22171
- Gene Name
- thymidylate synthase
- Gene Aliases
-
Ts
- Chromosome Location
-
Chr.5: 30058200 - 30073625 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033402.1 |
|
6 - 6 |
805 |
88 |
GenBank mRNA |
AK164981.1 |
- |
7 - 7 |
884 |
88 |
- Gene Symbol
- Tyms-ps
- Entrez Gene ID
-
22172
- Gene Name
- thymidylate synthase, pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 87966671 - 87968017 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_000040.2 |
|
1 - 1 |
894 |
88 |
View Full Product Details