Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsga10
Entrez Gene ID
211484
Gene Name
testis specific 10
Gene Aliases
4933432N21Rik, AI843383, Gm217, Mtsga10
Chromosome Location
Chr.1: 37728144 - 37866739 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290720.1 NP_001277649.1 7 - 8 1103 62
NM_001290721.1 NP_001277650.1 6 - 7 910 62
NM_207228.3 NP_997111.1 7 - 8 970 62
NR_110976.1 6 - 7 910 62
GenBank mRNA AF530050.3 - 6 - 7 915 62
AK030254.1 - 7 - 8 1103 62
AK163138.1 - 7 - 8 917 62
AK164954.1 - 7 - 8 937 62
AY618264.1 - 6 - 7 915 62
BC053407.1 - 6 - 7 893 62
BC066782.1 - 7 - 8 967 62

View Full Product Details