Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb38
Entrez Gene ID
245007
Gene Name
zinc finger and BTB domain containing 38
Gene Aliases
A930014K01Rik, CIBZ
Chromosome Location
Chr.9: 96685393 - 96731675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175537.3 NP_780746.2 1 - 2 388 72
GenBank mRNA AK044467.1 - 1 - 2 390 72

View Full Product Details