Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Taf1
Entrez Gene ID
270627
Gene Name
TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
AU015687, B430306D02Rik, Ccg-1, Ccg1, KAT4, N-TAF1, TAFII250, Taf2a, p250
Chromosome Location
Chr.X: 101527574 - 101601789 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290729.1 NP_001277658.1 25 - 26 4143 85
GenBank mRNA AB299229.1 - 25 - 26 4070 85
AK046668.1 - 4 - 5 632 85
BC047418.1 - 15 - 16 2256 85
BC094568.1 - 7 - 8 1427 85

View Full Product Details