The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111078.1 | NP_001104548.1 | 9 - 10 | 1410 | 68 |
NM_001111079.1 | NP_001104549.1 | 9 - 10 | 1530 | 68 | |
NM_001111080.1 | NP_001104550.1 | 9 - 10 | 1386 | 68 | |
NM_010931.3 | NP_035061.3 | 9 - 10 | 1554 | 68 | |
GenBank mRNA | AF274046.1 | - | 9 - 10 | 1467 | 68 |
AK143688.1 | - | 9 - 10 | 1379 | 68 | |
AK145376.1 | - | 9 - 10 | 1376 | 68 | |
AK145419.1 | - | 9 - 10 | 1376 | 68 | |
AK145543.1 | - | 9 - 10 | 1494 | 68 | |
AK145583.1 | - | 9 - 10 | 1376 | 68 | |
AK146951.1 | - | 9 - 10 | 1527 | 68 | |
AK147046.1 | - | 9 - 10 | 1470 | 68 | |
AK150489.1 | - | 9 - 10 | 1386 | 68 | |
AK151701.1 | - | 9 - 10 | 1376 | 68 | |
AK151837.1 | - | 9 - 10 | 1386 | 68 | |
AK152930.1 | - | 9 - 10 | 1386 | 68 | |
AK153083.1 | - | 9 - 10 | 1386 | 68 | |
BC022167.1 | - | 9 - 10 | 1482 | 68 | |
D87908.1 | - | 9 - 10 | 1465 | 68 |