Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trex1
Entrez Gene ID
22040
Gene Name
three prime repair exonuclease 1
Gene Aliases
1661, AU041952
Chromosome Location
Chr.9: 109057932 - 109059723 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC011133.1 - 1 - 2 82 108
BY234700.1 - 1 - 2 76 108

View Full Product Details