Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw21
Entrez Gene ID
320082
Gene Name
F-box and WD-40 domain protein 21
Gene Aliases
E330009P21Rik
Chromosome Location
Chr.9: 109136887 - 109162022 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177069.3 NP_796043.1 7 - 8 1023 81
GenBank mRNA AK087710.1 - 7 - 8 1024 81
AK135949.1 - 7 - 8 1024 81
AK136136.1 - 7 - 8 1020 81
BC099397.1 - 7 - 8 1005 81

View Full Product Details