Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zap70
Entrez Gene ID
22637
Gene Name
zeta-chain (TCR) associated protein kinase
Gene Aliases
Srk, ZAP-70, mrtle, mur
Chromosome Location
Chr.1: 36761798 - 36782821 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289765.1 NP_001276694.1 5 - 6 984 89
NM_001289766.1 NP_001276695.1 5 - 6 949 89
NM_009539.3 NP_033565.2 5 - 6 941 89
GenBank mRNA AK039883.1 - 5 - 6 878 89
AK089056.1 - 5 - 6 914 89
U77667.1 - 4 - 5 787 89

View Full Product Details