The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153408.2 | NP_700457.1 | 3 - 4 | 633 | 65 |
GenBank mRNA | AF321278.1 | - | 3 - 4 | 633 | 65 |
AK156207.1 | - | 3 - 4 | 644 | 65 | |
BC056622.1 | - | 3 - 4 | 624 | 65 |