Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Actr1b
Entrez Gene ID
226977
Gene Name
ARP1 actin-related protein 1B, centractin beta
Gene Aliases
2310066K23Rik, AA960180, AI851923, Arp1b
Chromosome Location
Chr.1: 36698114 - 36710017 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146107.2 NP_666219.1 9 - 10 1061 113
GenBank mRNA AK088839.1 - 9 - 10 1094 113
BC023020.1 - 9 - 10 1061 113
BC024861.1 - 4 - 5 385 113
BC027134.1 - - 1727 113

View Full Product Details