The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013785.2 | NP_038813.2 | 1 - 2 | 174 | 64 |
GenBank mRNA | AK037925.1 | - | 1 - 2 | 195 | 64 |
AK088872.1 | - | 1 - 2 | 191 | 64 | |
AK166143.1 | - | 1 - 2 | 178 | 64 | |
AK171310.1 | - | 1 - 2 | 179 | 64 | |
AK172921.1 | - | 1 - 2 | 233 | 64 | |
BC053030.1 | - | 1 - 2 | 180 | 64 | |
BC059006.1 | - | 1 - 2 | 174 | 64 |