Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Traip
Entrez Gene ID
22036
Gene Name
TRAF-interacting protein
Gene Aliases
Trip
Chromosome Location
Chr.9: 107950136 - 107972270 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011634.3 NP_035764.2 7 - 8 729 68
GenBank mRNA AK012786.1 - 7 - 8 711 68
AK012948.1 - 7 - 8 728 68
BC017374.1 - 7 - 8 718 68
U77844.1 - 7 - 8 734 68

View Full Product Details