Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rassf1
- Entrez Gene ID
-
56289
- Gene Name
- Ras association (RalGDS/AF-6) domain family member 1
- Gene Aliases
-
123F2, AA536941, AU044980, NORE2A, RDA32, REH3P21, Rassf1A, Rassf1B, Rassf1C
- Chromosome Location
-
Chr.9: 107551555 - 107562267 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF132851.1 |
- |
1 - 2 |
174 |
79 |
BC002173.1 |
- |
1 - 2 |
268 |
79 |
View Full Product Details