The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199217.1 | NP_001186146.1 | 11 - 12 | 1082 | 72 |
NM_001199218.1 | NP_001186147.1 | 11 - 12 | 1102 | 72 | |
NM_023805.3 | NP_076294.2 | 11 - 12 | 1250 | 72 | |
GenBank mRNA | AF159856.1 | - | 11 - 12 | 1052 | 72 |
AK002607.1 | - | 11 - 12 | 1068 | 72 | |
AK035155.1 | - | - | 1270 | 72 | |
BC010678.1 | - | 11 - 12 | 1082 | 72 | |
BC054846.1 | - | 11 - 12 | 1225 | 72 | |
BC055339.1 | - | 11 - 12 | 1102 | 72 |