Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mon1a
Entrez Gene ID
72825
Gene Name
MON1 homolog A (yeast)
Gene Aliases
2810468K17Rik
Chromosome Location
Chr.9: 107888112 - 107903139 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028369.3 NP_082645.1 5 - 6 1820 65
GenBank mRNA AK013387.1 - 5 - 6 1767 65
AK172624.1 - 5 - 6 1727 65
AK211446.1 - 2 - 3 252 65
BC006070.1 - 2 - 3 627 65
BC048852.1 - 5 - 6 1746 65

View Full Product Details