The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146092.2 | NP_666204.2 | 2 - 3 | 164 | 96 |
GenBank mRNA | AK144881.1 | - | 2 - 3 | 131 | 96 |
BC020299.1 | - | 2 - 3 | 137 | 96 | |
BC028647.1 | - | 2 - 3 | 111 | 96 |