Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp34
Entrez Gene ID
17847
Gene Name
ubiquitin specific peptidase 34
Gene Aliases
A530081C03Rik, Murr2
Chromosome Location
Chr.11: 23306846 - 23490560 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190401.2 NP_001177330.2 56 - 57 7156 135
GenBank mRNA AK129165.1 - 4 - 5 244 135
CN533811.1 - 2 - 3 118 135

View Full Product Details