Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dach1
Entrez Gene ID
13134
Gene Name
dachshund 1 (Drosophila)
Gene Aliases
Dac, Dach
Chromosome Location
Chr.14: 97786846 - 98169765 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038610.2 NP_001033699.1 9 - 10 2446 67
NM_007826.3 NP_031852.1 10 - 11 2602 67
GenBank mRNA AF090436.1 - 10 - 11 2146 67
AF090437.1 - 9 - 10 1990 67
AF102547.1 - 10 - 11 2146 67
AF129510.1 - 9 - 10 2208 67
AJ005669.1 - 10 - 11 1664 67
AK047409.1 - 9 - 10 1312 67
AK053594.1 - 9 - 10 1655 67
BC059934.1 - 9 - 10 2224 67
BC078644.1 - 9 - 10 1980 67
BC141130.1 - 9 - 10 2052 67
BC145440.1 - 9 - 10 2053 67

View Full Product Details