The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024560.2 | NP_001019731.1 | 7 - 8 | 832 | 66 |
GenBank mRNA | AK037118.1 | - | - | 823 | 66 |
AK047234.1 | - | 7 - 8 | 808 | 66 | |
AK143441.1 | - | 6 - 7 | 944 | 66 | |
BC048922.1 | - | 7 - 8 | 832 | 66 |