Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gstp2
Entrez Gene ID
14869
Gene Name
glutathione S-transferase, pi 2
Gene Aliases
GSTpiA, Gst-3, Gst3
Chromosome Location
Chr.19: 4040288 - 4042221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181796.2 NP_861461.1 1 - 2 57 79
GenBank mRNA BC064781.1 - 1 - 2 57 79
BC094623.1 - 1 - 2 61 79
BC100587.1 - 1 - 2 74 79

View Full Product Details