Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpc6
Entrez Gene ID
23888
Gene Name
glypican 6
Gene Aliases
-
Chromosome Location
Chr.14: 116921001 - 117979529 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079844.2 NP_001073313.1 3 - 4 1351 73
NM_011821.3 NP_035951.1 3 - 4 1351 73
GenBank mRNA AF105268.1 - 4 - 5 1074 73
AK032866.1 - 3 - 4 772 73
AK132531.1 - 3 - 4 1088 73
AK135442.1 - 3 - 4 1333 73
AK149312.1 - 3 - 4 1118 73
AK164668.1 - 3 - 4 1353 73
BC023448.1 - 3 - 4 765 73

View Full Product Details