Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zic4
Entrez Gene ID
22774
Gene Name
zinc finger protein of the cerebellum 4
Gene Aliases
-
Chromosome Location
Chr.9: 91361777 - 91389349 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009576.2 NP_033602.2 1 - 2 252 73
GenBank mRNA AK046913.1 - 1 - 2 253 73

View Full Product Details