Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdca5
Entrez Gene ID
67849
Gene Name
cell division cycle associated 5
Gene Aliases
2610036L13Rik, AL024086, AW536684, C85404
Chromosome Location
Chr.19: 6085097 - 6091773 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026410.3 NP_080686.1 5 - 6 778 75
GenBank mRNA AK010540.1 - 5 - 6 745 75
AK011701.1 - 5 - 6 778 75
AK016305.1 - 5 - 6 892 75
BC029626.1 - 4 - 5 672 75
BC052904.1 - 5 - 6 776 75

View Full Product Details