Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pla2g16
Entrez Gene ID
225845
Gene Name
phospholipase A2, group XVI
Gene Aliases
C78643, HRSL3, Hrasls3, Hrev107, MLP-3
Chromosome Location
Chr.19: 7557434 - 7588545 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK039703.1 - 1 - 2 320 95

View Full Product Details