Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snhg1
Entrez Gene ID
83673
Gene Name
small nucleolar RNA host gene 1
Gene Aliases
-
Chromosome Location
Chr.19: 8723487 - 8726326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002896.3 9 - 10 390 125
GenBank mRNA AK043166.1 - 9 - 10 391 125


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ178183.1 - 4 - 5 652 Chr. 19 - 8724663 - 8726299 [+] on Build GRCm38 125
GenBankmRNA AA646653.1 - 8 - 9 359 Chr. 19 - 8724152 - 8726312 [+] on Build GRCm38 125
GenBankmRNA AA259375.1 - 4 - 5 272 Chr. 19 - 8725107 - 8726417 [+] on Build GRCm38 125
GenBankmRNA AV497308.1 - 8 - 9 370 Chr. 19 - 8724155 - 8726331 [+] on Build GRCm38 125
GenBankmRNA BB816775.1 - 5 - 6 199 Chr. 19 - 8725011 - 8726441 [+] on Build GRCm38 125
GenBankmRNA AA433008.1 - 8 - 9 375 Chr. 19 - 8724140 - 8726432 [+] on Build GRCm38 126
GenBankmRNA AV460418.1 - 8 - 9 374 Chr. 19 - 8724138 - 8726323 [+] on Build GRCm38 125
GenBankmRNA EL960934.1 - 8 - 9 377 Chr. 19 - 8724140 - 8726443 [+] on Build GRCm38 125
GenBankmRNA BB640482.1 - 9 - 10 387 Chr. 19 - 8723490 - 8726328 [+] on Build GRCm38 125
GenBankmRNA CB589630.1 - 351 Chr. - - [-] on Build 125
GenBankmRNA BE448643.1 - 8 - 9 363 Chr. 19 - 8724148 - 8726344 [+] on Build GRCm38 125
GenBankmRNA C78065.1 - Chr. - - [-] on Build 125
GenBankmRNA DV053423.1 - 409 Chr. - - [-] on Build 125
GenBankmRNA AI661961.1 - 1 - 2 Chr. 19 - 8725023 - 8726443 [-] on Build GRCm38 125
GenBankmRNA BU898422.1 - 3 - 4 101 Chr. 19 - 8725506 - 8726326 [+] on Build GRCm38 125
GenBankmRNA BY098836.1 - 6 - 7 239 Chr. 19 - 8724673 - 8726359 [+] on Build GRCm38 125
GenBankmRNA AL034970.1 - 8 - 9 383 Chr. 19 - 8724138 - 8726409 [+] on Build GRCm38 125
GenBankmRNA AA939588.1 - 1 - 2 Chr. 19 - 8725011 - 8726443 [-] on Build GRCm38 125
GenBankmRNA AA408009.1 - 1 - 2 Chr. 19 - 8724640 - 8726440 [-] on Build GRCm38 125
GenBankmRNA AA795219.1 - 6 - 7 236 Chr. 19 - 8724676 - 8726370 [+] on Build GRCm38 125
GenBankmRNA AA178616.1 - 8 - 9 337 Chr. 19 - 8724174 - 8726395 [+] on Build GRCm38 125
GenBankmRNA CO043924.1 - 1 - 2 Chr. 19 - 8725158 - 8726443 [-] on Build GRCm38 125
GenBankmRNA BU757760.1 - 1 - 2 Chr. 19 - 8724897 - 8726443 [-] on Build GRCm38 125
GenBankmRNA CX567991.1 - 8 - 9 377 Chr. 19 - 8724140 - 8726443 [+] on Build GRCm38 125
GenBankmRNA AV570392.1 - 8 - 9 Chr. 19 - 8724157 - 8726390 [+] on Build GRCm38 125
GenBankmRNA W42022.1 - 369 Chr. - - [-] on Build 125
GenBankmRNA AW990131.1 - 8 - 9 362 Chr. 19 - 8724149 - 8726394 [+] on Build GRCm38 125
GenBankmRNA CR518605.1 - 1 - 2 Chr. 19 - 8725236 - 8726443 [-] on Build GRCm38 125
GenBankmRNA BM730535.1 - 8 - 9 357 Chr. 19 - 8724154 - 8726421 [+] on Build GRCm38 125
GenBankmRNA CX566533.1 - 8 - 9 505 Chr. 19 - 8724005 - 8726443 [+] on Build GRCm38 125
GenBankmRNA BG092552.1 - 1 - 2 Chr. 19 - 8725780 - 8726442 [-] on Build GRCm38 125
GenBankmRNA AV492597.1 - 8 - 9 376 Chr. 19 - 8724140 - 8726345 [+] on Build GRCm38 125
GenBankmRNA AA200918.1 - 6 - 7 245 Chr. 19 - 8724669 - 8726420 [+] on Build GRCm38 125
GenBankmRNA AV576327.1 - 3 - 4 205 Chr. 19 - 8725500 - 8726440 [+] on Build GRCm38 223
GenBankmRNA AV487936.1 - 8 - 9 341 Chr. 19 - 8724170 - 8726374 [+] on Build GRCm38 125
GenBankmRNA AV481080.1 - 8 - 9 354 Chr. 19 - 8724157 - 8726376 [+] on Build GRCm38 125
GenBankmRNA AV474095.1 - 3 - 4 162 Chr. 19 - 8725444 - 8726443 [+] on Build GRCm38 125
GenBankmRNA AV569303.1 - 8 - 9 347 Chr. 19 - 8724164 - 8726438 [+] on Build GRCm38 125
GenBankmRNA BE849804.1 - 3 - 4 123 Chr. 19 - 8725484 - 8726443 [+] on Build GRCm38 125
GenBankmRNA BQ268643.1 - 1 - 2 Chr. 19 - 8724444 - 8726443 [-] on Build GRCm38 125
GenBankmRNA AA028518.1 - 7 - 8 289 Chr. 19 - 8724435 - 8726342 [+] on Build GRCm38 125
GenBankmRNA AA543872.1 - 6 - 7 234 Chr. 19 - 8724678 - 8726442 [+] on Build GRCm38 125
GenBankmRNA BQ416548.1 - 8 - 9 365 Chr. 19 - 8724146 - 8726443 [+] on Build GRCm38 125
GenBankmRNA BX525735.1 - 8 - 9 364 Chr. 19 - 8724146 - 8726398 [+] on Build GRCm38 125
GenBankmRNA AV445236.1 - 8 - 9 380 Chr. 19 - 8724157 - 8726345 [+] on Build GRCm38 125
GenBankmRNA AA220561.1 - 8 - 9 352 Chr. 19 - 8724159 - 8726387 [+] on Build GRCm38 125
GenBankmRNA EL962085.1 - 8 - 9 424 Chr. 19 - 8724140 - 8726432 [+] on Build GRCm38 125
GenBankmRNA BG090562.1 - 8 - 9 334 Chr. 19 - 8724177 - 8726313 [+] on Build GRCm38 125
GenBankmRNA AI155059.1 - 3 - 4 344 Chr. 19 - 8725261 - 8726385 [+] on Build GRCm38 125
GenBankmRNA AW985636.1 - 8 - 9 373 Chr. 19 - 8724140 - 8726383 [+] on Build GRCm38 125
GenBankmRNA BX528587.1 - 8 - 9 375 Chr. 19 - 8724140 - 8726387 [+] on Build GRCm38 125
GenBankmRNA AV495678.1 - 4 - 5 272 Chr. 19 - 8725108 - 8726443 [+] on Build GRCm38 125
GenBankmRNA AV584788.1 - 7 - 8 321 Chr. 19 - 8724418 - 8726393 [+] on Build GRCm38 125
GenBankmRNA CF896937.1 - 8 - 9 363 Chr. 19 - 8724148 - 8726442 [+] on Build GRCm38 125
GenBankmRNA AV473637.1 - 5 - 6 178 Chr. 19 - 8725029 - 8726443 [+] on Build GRCm38 125
GenBankmRNA BI068068.1 - 3 - 4 303 Chr. 19 - 8725304 - 8726388 [+] on Build GRCm38 125
GenBankmRNA AV505845.1 - 8 - 9 391 Chr. 19 - 8724147 - 8726323 [+] on Build GRCm38 125
GenBankmRNA AV579856.1 - 3 - 4 121 Chr. 19 - 8725486 - 8726443 [+] on Build GRCm38 125
GenBankmRNA AA462480.1 - 8 - 9 374 Chr. 19 - 8724140 - 8726311 [+] on Build GRCm38 125
GenBankmRNA AV469387.1 - 6 - 7 210 Chr. 19 - 8724704 - 8726443 [+] on Build GRCm38 125
GenBankmRNA BU898430.1 - 3 - 4 101 Chr. 19 - 8725506 - 8726326 [+] on Build GRCm38 125
GenBankmRNA AV481116.1 - 8 - 9 353 Chr. 19 - 8724158 - 8726391 [+] on Build GRCm38 125
GenBankmRNA BF608050.1 - 8 - 9 375 Chr. 19 - 8724147 - 8726443 [+] on Build GRCm38 125
GenBankmRNA BM730662.1 - 1 - 2 Chr. 19 - 8724156 - 8726443 [-] on Build GRCm38 125
GenBankmRNA BQ177137.1 - 1 - 2 Chr. 19 - 8723475 - 8726443 [-] on Build GRCm38 125
GenBankmRNA BG092665.1 - 1 - 2 Chr. 19 - 8724429 - 8726443 [-] on Build GRCm38 125
GenBankmRNA BP427963.1 - 6 - 7 Chr. 19 - 8724669 - 8726443 [+] on Build GRCm38 125
GenBankmRNA AV501497.1 - 8 - 9 354 Chr. 19 - 8724157 - 8726389 [+] on Build GRCm38 125
GenBankmRNA BB254596.2 - 9 - 10 430 Chr. 19 - 8723487 - 8726326 [+] on Build GRCm38 125
GenBankmRNA AA760260.1 - 7 - 8 307 Chr. 19 - 8724140 - 8726396 [+] on Build GRCm38 125


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nucleolar RNA, C/D box 22 Snord22 Rnu22, U22 -76.0 Sense Chr. - 8725866 - 8725991 on Build GRCm38
Downstream WD repeat domain 74 Wdr74 5730436H21Rik, AA407588 9.4kb Sense Chr. - 8735799 - 8740624 on Build GRCm38

View Full Product Details