Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cep70
Entrez Gene ID
68121
Gene Name
centrosomal protein 70
Gene Aliases
6720484E09Rik, Bite, C030018L16Rik
Chromosome Location
Chr.9: 99183595 - 99302065 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023873.3 NP_076362.2 9 - 10 951 105
GenBank mRNA AK133048.1 - 9 - 10 957 105
BC050819.1 - 9 - 10 943 105
BC071260.1 - 9 - 10 951 105

View Full Product Details