The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026763.2 | NP_081039.2 | 16 - 17 | 4525 | 85 |
GenBank mRNA | AB370265.1 | - | 16 - 17 | 4617 | 85 |
AM231153.1 | - | 10 - 11 | 1905 | 85 |