Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Traip
Entrez Gene ID
22036
Gene Name
TRAF-interacting protein
Gene Aliases
Trip
Chromosome Location
Chr.9: 107950136 - 107972270 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC006929.1 - 1 - 2 246 68

View Full Product Details