Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Yipf4
Entrez Gene ID
67864
Gene Name
Yip1 domain family, member 4
Gene Aliases
2310034L04Rik
Chromosome Location
Chr.17: 74489493 - 74500277 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC003936.1 - 1 - 2 244 128

View Full Product Details