Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Strn
Entrez Gene ID
268980
Gene Name
striatin, calmodulin binding protein
Gene Aliases
AU022939, D130055P19
Chromosome Location
Chr.17: 78649908 - 78737191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011500.2 NP_035630.2 12 - 13 1625 103
GenBank mRNA AJ223777.1 - 12 - 13 1552 103
AK135684.1 - 7 - 8 818 103
AK136032.1 - 10 - 11 1274 103
BC090968.1 - 12 - 13 1625 103
BC150727.1 - 12 - 13 1593 103

View Full Product Details