The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028721.3 | NP_082997.3 | 2 - 3 | 742 | 104 |
NM_172460.1 | NP_766048.1 | 2 - 3 | 742 | 104 | |
GenBank mRNA | AK048877.1 | - | 2 - 3 | 515 | 104 |
AK052281.1 | - | 2 - 3 | 529 | 104 | |
AK133217.1 | - | 2 - 3 | 743 | 104 | |
AK173327.1 | - | 2 - 3 | 321 | 104 | |
AY259499.1 | - | 2 - 3 | 682 | 104 | |
BC115724.1 | - | 2 - 3 | 272 | 104 |