Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chic1
Entrez Gene ID
12212
Gene Name
cysteine-rich hydrophobic domain 1
Gene Aliases
Brx
Chromosome Location
Chr.X: 103356328 - 103409096 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009767.2 NP_033897.1 2 - 3 473 67
GenBank mRNA AK034427.1 - 2 - 3 446 67
AK135974.1 - 2 - 3 425 67
BC132322.1 - 2 - 3 392 67
BC138730.1 - 2 - 3 392 67

View Full Product Details