The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166474.1 | NP_001159946.1 | 1 - 2 | 104 | 83 |
GenBank mRNA | BC039944.1 | - | 1 - 2 | 68 | 83 |
BC058424.1 | - | 1 - 2 | 85 | 83 |