The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252466.1 | NP_001239395.1 | 14 - 15 | 1467 | 78 |
NM_001252467.1 | NP_001239396.1 | 11 - 12 | 1107 | 78 | |
NM_027933.3 | NP_082209.1 | 13 - 14 | 1266 | 78 | |
NR_045519.1 | 14 - 15 | 1376 | 78 | ||
GenBank mRNA | AK011541.1 | - | 6 - 7 | 627 | 78 |
AK133949.1 | - | 13 - 14 | 1113 | 78 | |
AK155301.1 | - | 13 - 14 | 1135 | 78 | |
AK167731.1 | - | 13 - 14 | 1116 | 78 | |
BC066005.1 | - | 13 - 14 | 1107 | 78 | |
BC145892.1 | - | 13 - 14 | 1130 | 78 | |
BC145894.1 | - | 13 - 14 | 1130 | 78 |