The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207515.2 | NP_997398.1 | 5 - 6 | 1485 | 119 |
GenBank mRNA | AK028797.1 | - | 5 - 6 | 1450 | 119 |
AK151913.1 | - | 5 - 6 | 1449 | 119 | |
AK151926.1 | - | 5 - 6 | 1449 | 119 | |
AK151957.1 | - | 5 - 6 | 1449 | 119 | |
AK159213.1 | - | 5 - 6 | 1443 | 119 | |
AK167479.1 | - | 5 - 6 | 1315 | 119 | |
AK167694.1 | - | 6 - 7 | 1250 | 119 | |
GQ906534.1 | - | 4 - 5 | 803 | 119 |