Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqcf2
Entrez Gene ID
547049
Gene Name
IQ motif containing F2
Gene Aliases
EG547049
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CN834216.1 - - 174 115

View Full Product Details